Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylammonium bromide, 99+%

Catalog No. AC220712500
Click to view available options
Quantity:
50 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle

Catalog No. AC220712500

Supplier: Thermo Scientific Chemicals 220712500

Only null left
Add to Cart

CAS: 71-91-0 | C8H20BrN | 210.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-91-0
Molecular Formula C8H20BrN
Molecular Weight (g/mol) 210.16
InChI Key HWCKGOZZJDHMNC-UHFFFAOYSA-M
Synonym tetraethylammonium bromide, teab, tea bromide, tetrylammonium bromide, sympatektoman, tetranium, etylon, bromethyl, beparon, etambro
PubChem CID 6285
IUPAC Name tetraethylazanium;bromide
SMILES CC[N+](CC)(CC)CC.[Br-]

Specifications

Melting Point 285.0°C
Color White to Cream
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C2H5)4NBr
Quantity 250 g
Beilstein 04,104
Fieser 06,568; 13,46
Merck Index 15,9344
Solubility Information Solubility in water: 2795g/L (25°C). Other solubilities: freely soluble in alcohol, chloroform and acetone, slightly soluble in benzene
Formula Weight 210.16
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tetraethylammonium bromide
Show More Show Less
EINECSNumber 200-769-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.