Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead(II,IV) oxide, red, 98%
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Melting Point | 500.0°C |
| Color | Orange |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 97.5% min. (PB3O4) |
| Linear Formula | 2PbO·PbO2 |
| Merck Index | 15,548 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: partly soluble in acids, insoluble in alcohol |
| Formula Weight | 685.57 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes damage to organs through prolonged or repeated exposure. Harmful if inhaled. Harmful if swallowed. May damage the unborn child. Suspected of damaging fertility. Suspected of causing cancer. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face,hands and any exposed skin thoroughly after handling. IF INHALED: Remove to fresh air and k |
| EINECSNumber | 215-235-6 |
| RTECSNumber | OG5425000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction