Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aniline, 99.5%, extra pure
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -6.2°C |
| Color | Brown to Colorless |
| Density | 1.0213g/mL |
| Boiling Point | 181.0°C to 185.0°C |
| Flash Point | 76°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99.4% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5845 to 1.5865 |
| Linear Formula | C6H5NH2 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Suspected of causing cancer. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Toxic in contact with skin. Toxic if swallowed. Causes serious eye dam |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF |
| EINECSNumber | 200-539-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction