Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenol, 99.5%, extra pure, loose crystals, unstabilized
Click to view available options
Quantity:
250 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Acidity | (per Eur. Pharm.) Passes Test |
| Melting Point | 39.0°C to 42.0°C |
| Color | Colorless to White |
| Density | 1.0700g/mL |
| Boiling Point | 182.0°C |
| Flash Point | 79°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | C6H5OH |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. Toxic if inhaled. Suspected of causing genetic defects. May cause damage to organs through prolonged or repeated |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and kee |
| EINECSNumber | 203-632-7 |
| RTECSNumber | SJ3325000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction