Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carnauba wax, refined, No. 1, yellow

Catalog No. AC222380050
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum

CAS: 8015-86-9

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8015-86-9
MDL Number MFCD00130724
Synonym Brazil wax

Specifications

Acidity 2 to 7 (mg KOH/g)
Melting Point 81.0°C to 86.0°C
Color Yellow
Flash Point 282°C
Ignition Residue 0.25% max.
Packaging Plastic drum
Quantity 5 kg
Saponification Value 78 to 95mg KOH/g
Merck Index 15,1846
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in boiling alcohol,diethyl ether,benzene,,toluene,chloroform,ethyl acetate,turpentine,,petroleum ether,carbon disulfide;,soluble in alkalies
Physical Form Powder or Flakes
Chemical Name or Material Carnauba wax
Show More Show Less
EINECSNumber 232-399-4
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.