Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromide, 99+%, for analysis

Catalog No. AC222550025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 7758-02-3 | BrK | 119.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435.0°C
Melting Point 734.0°C
Color White
pH 5.0 to 8.8 (5% soln. at 20°C)
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 2.5 kg
Assay Percent Range 99% min. (Argentometry)
Linear Formula KBr
Merck Index 15, 7739
Solubility Information Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C),1.42g/L ethanol (25°C),217g/L glycerol,4g/L alcohol
IUPAC Name potassium;bromide
Formula Weight 119
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.2% max. (130°C)
Packaging Plastic bottle
Chemical Name or Material Potassium bromide, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 231-830-3
RTECSNumber TS7650000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.