Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) n-butoxide, 98+%

Catalog No. AC223190010
Click to view available options
Quantity:
250 g
500 g
1 kg
2 kg
10 kg
Packaging:
Glass bottle
Plastic Drum

CAS: 5593-70-4 | C16H36O4Ti | 340.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5593-70-4
Molecular Formula C16H36O4Ti
Molecular Weight (g/mol) 340.35
MDL Number MFCD00009433
InChI Key FPCJKVGGYOAWIZ-UHFFFAOYSA-N
Synonym butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
PubChem CID 25203927
IUPAC Name butan-1-ol;titanium
SMILES CCCCO.CCCCO.CCCCO.CCCCO.[Ti]

Specifications

Melting Point -75°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 310.0°C to 314.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.486
Linear Formula Ti[O(CH2)3CH3]4
Quantity 1 kg
Specific Gravity 0.99
Solubility Information Solubility in water: rapidly hydrolized by water. Other solubilities: miscible with most common organic solvents
Viscosity 50-80 mPa.s (20°C)
Formula Weight 340.35
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Titanium(IV) n-butoxide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at
EINECSNumber 227-006-8
RTECSNumber XR1585000
TSCA TSCA
Recommended Storage the product darkens with age

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.