Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chlorate, 99+%, extra pure

Catalog No. AC223222500
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle

Catalog No. AC223222500

Supplier: Thermo Scientific Chemicals 223222500

Only null left
Add to Cart

CAS: 7775-09-9 | ClNaO3 | 106.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-09-9
Molecular Formula ClNaO3
Molecular Weight (g/mol) 106.44
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
Synonym sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
PubChem CID 516902
ChEBI CHEBI:65242
SMILES [Na+].[O-][Cl](=O)=O

Specifications

Melting Point 248.0°C to 261.0°C
Color White
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99+%
Linear Formula NaClO3
Fieser 01,1056
Merck Index 15,8733
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and glycerin,7.7g/L ethanol (20°C)
IUPAC Name sodium;chlorate
Formula Weight 106.44
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 2.49
Density 2.4900g/mL
Chemical Name or Material Sodium chlorate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
May cause fire or explosion; strong oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 231-887-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.