Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethanol, for spectroscopy, anhydrous, denat. with 5% 2-Propanol + 5% Methanol
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for UV/VIS spectroscopy
Specifications
Specifications
| Absorbance | 0.01 max. at 300nm, 0.04 max. at 250nm, 0.65 max. at 210nm, 1 max. at 205nm |
| Density | 1.59 |
| Quantity | 500 mL |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Residue after Evaporation | 0.0005% max. |
| Linear Formula | C2H5OH |
| Vapor Pressure | Vapor Pressure: 59mbar at 20°C |
| Water | 0.1% Max. (Coulometric) |
| Merck Index | 14, 3761 |
| Show More |
For Manufacturing and Laboratory Use. Not for beverage use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction