Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-Styrene oxide, 95%, 95% ee

Catalog No. AC240550050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 20780-53-4 | C8H8O | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20780-53-4
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00066210
InChI Key AWMVMTVKBNGEAK-QMMMGPOBSA-N
Synonym r-styrene oxide, r-phenyloxirane, r-+-styrene oxide, r-2-phenyloxirane, 2r-2-phenyloxirane, r-epoxyethyl benzene, oxirane, phenyl-, 2r, benzene, epoxyethyl-, r
PubChem CID 114705
ChEBI CHEBI:45389
IUPAC Name (2R)-2-phenyloxirane
SMILES C1O[C@@H]1C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -37.0°C
Color Yellow
Density 1.0530g/mL
Boiling Point 192.0°C to 195.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5360
Quantity 5 g
Specific Gravity 1.053
Specific Rotation Condition −21° (23°C c=5,chloroform)
Specific Rotation − 21.00
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in hexane,petroleum ether,acetone,,alcohol,and benzene
Formula Weight 120.15
Percent Purity 95%,95% ee
Physical Form Liquid
Chemical Name or Material (R)-Styrene oxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
May cause genetic defects.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.