Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloroperoxybenzoic Acid, 70-75%, balance 3-Chlorobenzoic Acid and water

Catalog No. AC255795000
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Plastic bottle

3-Chloroperoxybenzoic acid, 70-75%, C7H5ClO3, CAS Number-937-14-4, 535-80-8, 7732-18-5 | CAS: 937-14-4 | C7H5ClO3 | 172.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Chloroperoxybenzoic Acid
Name Note 70 - 75%
CAS 937-14-4, 535-80-8, 7732-18-5
CAS Min % 70.0, 25.0, 25.0
CAS Max % 75.0, 30.0, 30.0
Melting Point 92.0°C to 94.0°C
Color White
Density 0.5600g/mL
Assay Percent Range di-m-Chlorobenzoyl peroxide: < 0.1% Typical (HPLC)
Packaging Plastic bottle
Molecular Formula C7H5ClO3
Linear Formula ClC6H4CO3H
Quantity 500 g
MDL Number MFCD00002127
UN Number 3106
Beilstein 09,IV,972
Fieser 01,135; 02,68; 03,49; 04,85; 05,120; 06,110; 07,62; 08,97; 09,108; 10,92; 11,122; 12,118; 13,76; 15,86; 16,80; 17,76
Merck Index 15,2154
Specific Gravity 0.56
Synonym 3-chloroperoxybenzoic acid, 3-chloroperbenzoic acid, mcpba, m-chloroperbenzoic acid, m-chloroperoxybenzoic acid, meta-chloroperoxybenzoic acid, benzenecarboperoxoic acid, 3-chloro, m-chlorobenzoyl hydroperoxide, meta-chloroperbenzoic acid, 3-chlorobenzoperoxoic acid
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in koh, soluble in chloroform and acetone
InChI Key NHQDETIJWKXCTC-UHFFFAOYSA-N
SMILES OOC(=O)C1=CC=CC(Cl)=C1
IUPAC Name 3-chlorobenzenecarboperoxoic acid
Molecular Weight (g/mol) 172.56
PubChem CID 70297
ChEBI CHEBI:52091
Formula Weight 172.57
Percent Purity 70-75%
Physical Form Moist Powder
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.