Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide dimethyl acetal, 97%, stabilized

Catalog No. AC264131000
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 18871-66-4 | C6H15NO2 | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18871-66-4
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008476
InChI Key FBZVZUSVGKOWHG-UHFFFAOYSA-N
Synonym n,n-dimethylacetamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxy-ethyl-dimethyl-amine, ethanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylacetamide dimethylacetal, dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332
PubChem CID 87835
IUPAC Name 1,1-dimethoxy-N,N-dimethylethanamine
SMILES CC(N(C)C)(OC)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9110g/mL
Boiling Point 110.0°C to 118.0°C
Flash Point 8°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4160 to 1.4180 (20°C, 589nm)
Quantity 100 g
Fieser 04,166; 05,226; 10,154
Specific Gravity 0.911
Stabilizer sodium carbonate
Formula Weight 133.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N,N-Dimethylacetamide dimethyl acetal
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and eas
EINECSNumber 242-641-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.