Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate, 85.0%, mixture of isomers, Thermo Scientific Chemicals

Catalog No. AC269012500
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19437-42-4
Molecular Formula C10H5Na3O9S3
Molecular Weight (g/mol) 434.29
MDL Number MFCD00149263 MFCD00003989
InChI Key NJPKYOIXTSGVAN-UHFFFAOYSA-K
Synonym 1,3,6-naphthalenetrisulfonic acid, sodium salt, 1,3,6-naphthalenetrisulfonic acid, sodium salt 1:?
PubChem CID 73555608
IUPAC Name sodium;5,7-disulfonaphthalene-2-sulfonate
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=CC(=CC(=C2C=C1)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Color Beige or White to Yellow
Loss on Drying 8% max.
Packaging Glass bottle
Quantity 250 g
Beilstein 11,229
Solubility Information Solubility in water: 600g/L (20°C)
Formula Weight 368.36
Percent Purity ≥90% (by difference)
Physical Form Powder
Chemical Name or Material 1,3,6-Naphthalenetrisulfonic acid, sodium salt hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 243-068-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.