Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™

Catalog No. AC277858000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 1070-89-9 | C6H18NNaSi2 | 183.377 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium bis(trimethylsilyl)amide
Color Orange to Brown
Name Note 2M Solution in THF
CAS 1070-89-9, 109-99-9
Molecular Formula C6H18NNaSi2
Linear Formula [(CH3)3Si]2NNa
MDL Number MFCD00009835
Fieser 01,1046; 03,261; 04,442; 06,529; 12,441; 16,307
Density 0.9160g/mL
Synonym sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
InChI Key WRIKHQLVHPKCJU-UHFFFAOYSA-N
SMILES C[Si](C)(C)[N-][Si](C)(C)C.[Na+]
IUPAC Name sodium;bis(trimethylsilyl)azanide
Molecular Weight (g/mol) 183.377
PubChem CID 2724254
Formula Weight 183.38
Percent Purity 38 to 42%
Grade Pure
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.916
Quantity 800 mL
Flash Point −21°C
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.