Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyridine, 99+%, for analysis
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -42.0°C |
| Density | 0.9780g/mL |
| Boiling Point | 115.0°C to 116.0°C |
| Flash Point | 17°C |
| Assay Percent Range | 99% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5085 to 1.5105 |
| Quantity | 2.5 L |
| Beilstein | 20,181 |
| Fieser | 01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful in contact with skin. Harmful if inhaled. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. |
| EINECSNumber | 203-809-9 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction