Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-(2-furyl)phosphine, 98%

Catalog No. AC294800010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

CAS: 5518-52-5 | C12H9O3P | 232.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5518-52-5
Molecular Formula C12H9O3P
Molecular Weight (g/mol) 232.18
MDL Number MFCD00151857
InChI Key DLQYXUGCCKQSRJ-UHFFFAOYSA-N
Synonym tri 2-furyl phosphine, tri furan-2-yl phosphine, tri-2-furyl phosphine, tris furan-2-yl phosphane, tri-2-furylphosphine, tris 2-furyl phosphine, tri furan-2-yl phosphane, trifurylphosphine, tri fur-2-yl phosphane, tris 2-furanyl phosphine
PubChem CID 521585
IUPAC Name tris(furan-2-yl)phosphane
SMILES O1C=CC=C1P(C1=CC=CO1)C1=CC=CO1

Specifications

Melting Point 61.0°C to 64.0°C
Boiling Point 136.0°C (4.0 mmHg)
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 232.18
Percent Purity 98%
Chemical Name or Material Tri-(2-furyl)phosphine, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage Refrigerator +4°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.