Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals

Catalog No. AC295522500
Click to view available options
Quantity:
250 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Acidity 0.04mg KOH/g max.
Density 1.1000g/mL
Boiling Point 189.0°C
Quantity 250 mL
Flash Point 87°C
Packaging Glass Bottle
Physical Form Liquid at 20°C
Refractive Index 1.4770 to 1.4800
Residue after Evaporation 0.003% max.
Assay Percent Range 99.8+%
Specific Gravity 1.1
Linear Formula (CH3)2SO
Viscosity 1.987 mPa.s (25°C)
Beilstein 01, IV, 1277
Water 0.1% max.
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Merck Index 15, 3285
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Formula Weight 78.13
Percent Purity ≥99.8%
Grade Extra Pure
Chemical Name or Material Methyl sulfoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.