Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Zinc acetate dihydrate, 98+%, ACS reagent
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                500 g
                            
                        
                            
                                2.5 kg
                            
                        
                            
                                10 kg
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Plastic bottle
                            
                        
                            
                                Plastic drum
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
            Specifications
Specifications
| Melting Point | 237.0°C | 
| Color | White | 
| pH | 6.0 to 7.0 (5% soln. at 25°C) | 
| Physical Form | Crystals | 
| Quantity | 500 g | 
| Assay Percent Range | 98.0 to 101% (Complexometry) | 
| Linear Formula | (CH3CO2)2Zn·2H2O | 
| Merck Index | 15,10327 | 
| Solubility Information | Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol | 
| Formula Weight | 219.5 | 
| Show More | 
Safety and Handling
| Hazard Category | GHS Signal Word: Warning | 
| Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful if swallowed. Very toxic to aquatic life. | 
| Precautionary Statement | GHS P Statement Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously wit | 
| RTECSNumber | ZG8750000 | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                