Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexyne, 98%

Catalog No. AC329901000
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 693-02-7 | C6H10 | 82.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-02-7
Molecular Formula C6H10
Molecular Weight (g/mol) 82.13
InChI Key CGHIBGNXEGJPQZ-UHFFFAOYSA-N
Synonym 1-hexyne, n-butylacetylene, butylacetylene, hexyne, unii-5fzf2f38f5, n-hexyne, 1-hexyn, hex-5-yn-1-yl, pubchem13057
PubChem CID 12732
IUPAC Name hex-1-yne
SMILES CCCCC#C

Specifications

Melting Point -132.0°C
Color Colorless
Density 0.7100g/mL
Boiling Point 71.0°C to 72.0°C
Flash Point −021°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3974
Linear Formula CH3(CH2)3C≡CH
Quantity 100 mL
Beilstein 01,253
Specific Gravity 0.71
Formula Weight 82.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Hexyne
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 211-736-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.