Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™

Catalog No. AC348435000
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle

N,N-Dimethylformamide, 99.8%, C3H7NO, CAS Number-68-12-2, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dwumetyloformamid, dimethylformamide, dmfa, dimethyl formamide, dimetilformamide, dmf, dimethylformamid, 1L, 0.01 % max., Vapor Pressure: 4.9mbar at 20 deg.C, 04, 58 | CAS: 68-12-2 | C3H7NO | 73.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61.0°C
Color White to Beige
CAS Min % 99.75
CAS Max % 100.0
Decomposition Information >350°C
Density 0.9450g/mL
Boiling Point 153.0°C
Quantity 500 mL
Flash Point 58°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Crystalline Powder
Refractive Index 1.4290 to 1.4320
Assay Percent Range 99.8%
Specific Gravity 0.945
Linear Formula HCON(CH3)2
Vapor Pressure Vapor Pressure: 4.9mbar at 20°C
Viscosity 0.8 mPa.s (20°C)
Beilstein 04, 58
Water 0.005% Max. (Coulometric)
Fieser 01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
Merck Index 15, 3267
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Formula Weight 73.09
Percent Purity 99.8%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material N,N-Dimethylformamide, water <50 ppm
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.