Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene
Click to view available options
Quantity:
2.5 L
10 L
Packaging:
Glass Bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For peptide synthesis

Specifications
Specifications
Melting Point | -97.0°C |
Density | 1.3250g/mL |
Boiling Point | 39.0°C to 40.0°C |
Quantity | 10 L |
Packaging | Metal Drum |
Physical Form | Liquid |
Assay Percent Range | 99.9% |
Specific Gravity | 1.325 |
Linear Formula | CH2Cl2 |
Viscosity | 0.43 mPa.s (20°C) |
Show More |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction