Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™
Click to view available options
Quantity:
100 mL
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -110.0°C |
Color | Colorless |
CAS Min % | 99 |
CAS Max % | 100 |
Density | 0.7404g/mL |
Boiling Point | 54.0°C to 56.0°C |
Quantity | 100 mL |
Flash Point | −28°C |
Infrared Spectrum | Authentic |
Physical Form | Liquid |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | GHS Signal Word: Danger |
missing translation for 'healthHazard2' | GHS H Statement Causes skin irritation. Highly flammable liquid and vapor. |
missing translation for 'healthHazard3' | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. |
EINECSNumber | 216-653-1 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction