Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl methyl ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™

Catalog No. AC375220010
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle

Catalog No. AC375220010

Supplier: Thermo Scientific Chemicals 375220010

Only null left
Add to Cart

CAS: 1634-04-4 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -110.0°C
Color Colorless
CAS Min % 99
CAS Max % 100
Density 0.7404g/mL
Boiling Point 54.0°C to 56.0°C
Quantity 1 L
Flash Point −28°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3675 to 1.3695
Assay Percent Range 99% min. (GC)
Specific Gravity 0.7404
Linear Formula (CH3)3COCH3
Viscosity 0.36 mPa.s (20°C)
Water 0.005% max.
Merck Index 15, 6106
Solubility Information Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
Formula Weight 88.15
Percent Purity 99+%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material tert-Butyl methyl ether, Over Molecular Sieves
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
EINECSNumber 216-653-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.