Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, 99+%, extra pure, stabilized with amylene

Catalog No. AC383760025
Click to view available options
Quantity:
1 L
2.5 L
25 L

CAS: 67-66-3 | CHCl3 | 119.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3, 513-35-9
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES ClC(Cl)Cl

Specifications

Melting Point -63.0°C
Density 1.4800g/mL
Boiling Point 61.0°C
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range 99+%
Specific Gravity 1.48
Linear Formula CHCl3
Viscosity 0.56 mPa.s (20°C)
Water 0.1% max. (K.F.)
Merck Index 15, 2142
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone.
Formula Weight 119.38
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Chloroform, Stabilized with amylene
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.
Precautionary Statement GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.