Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid, triammonium salt, 97+%, pure

Catalog No. AC391410010
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle

CAS: 3458-72-8 | C6H20N3O7+3 | 246.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-72-8
Molecular Formula C6H20N3O7+3
Molecular Weight (g/mol) 246.24
MDL Number MFCD00036406
InChI Key YWYZEGXAUVWDED-UHFFFAOYSA-Q
Synonym ammonium citrate
PubChem CID 131675891
IUPAC Name azane;[3-carboxy-1,3-dihydroxy-4-[oxonio(oxoniumylidene)methyl]butylidene]oxidanium
SMILES C(C(=[OH+])[OH2+])C(CC(=[OH+])O)(C(=O)O)O.N.N.N

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Plastic bottle
Quantity 1 kg
Formula Weight 243.22
Grade Extra Pure
Physical Form Crystals
Chemical Name or Material Citric acid, triammonium salt
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.