Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™

Catalog No. AC396620010
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog No. Quantity Packaging
AC396620010 1 L AcroSeal™ Glass Bottle
AC396621000 100 mL AcroSeal™ Glass Bottle
AC396620025 2.5 L AcroSeal™ Glass Bottle
3 options

Catalog No. AC396620010

Supplier: Thermo Scientific Chemicals 396620010

Only null left
Add to Cart

CAS: 96-47-9 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-47-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00005367
InChI Key JWUJQDFVADABEY-UHFFFAOYSA-N
Synonym 2-methyltetrahydrofuran, methyltetrahydrofuran, tetrahydrosylvan, tetrahydro-2-methylfuran, furan, tetrahydro-2-methyl, furan, 2-methyl-tetrahydro, 2-methyl tetrahydrofuran, 2-methyl-tetrahydrofuran, 2-methyl-tetrahydro-furan, methyl tetrahydrofuran
PubChem CID 7301
IUPAC Name 2-methyloxolane
SMILES CC1CCCO1

Specifications

Melting Point -136.0°C
Color Colorless
CAS Min % 99.0
CAS Max % 100.0
Density 0.8600g/mL
Boiling Point 78.0°C to 80.0°C
Quantity 1 L
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.4046 to 1.4066
Assay Percent Range 99% min. (GC)
Specific Gravity 0.86
Viscosity 4 mPa.s (25°C)
Merck Index 15,6198
Solubility Information Solubility in water: 150g/l (25°C)
Formula Weight 86.13
Percent Purity 99+%
Grade Extra Dry
Chemical Name or Material 2-Methyltetrahydrofuran, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
EINECSNumber 202-507-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.