Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclohexanone, 99+%, ACS reagent
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -47.0°C |
Density | 0.9470g/mL |
Boiling Point | 155.0°C |
Quantity | 1 L |
Flash Point | 43°C |
Refractive Index | 1.4450 to 1.4560 |
Specific Gravity | 0.947 |
Linear Formula | C6H10(=O) |
Water | 0.05% max. |
Solubility Information | Solubility in water: 150g/L (10°C). Other solubilities: soluble in all common organic solvents |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement Flammable liquid and vapor. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye damage. Causes skin irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction