Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur.

Catalog No. AC408820025
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic Drum

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Melting Point 12.0°C
CAS Min % 99.0
CAS Max % 100.0
Density 1.0340g/mL
Boiling Point 101.0°C
Quantity 2.5 L
Flash Point 12°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4205 to 1.4220
Residue after Evaporation 0.005% max.
Assay Percent Range 99+%
Specific Gravity 1.034
Titratable Acid 0.0016 meq/g max.
Viscosity 1.32 mPa.s (20°C)
Beilstein 19, 3
Water 0.05% max. (Coulometric)
Fieser 01,333
Merck Index 15, 3328
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
Formula Weight 88.11
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material 1, 4-Dioxane
Show More Show Less
Hazard Category SIGNAL WORD: DANGER!
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May form explosive peroxides. Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. Inhalation may cause central nervous system effects. Repeated exposure may cause skin dryness or cracking. Hygroscopic.
NFPA
Health: 2
Flammability: 3
Instability: 1
EINECSNumber 204-661-8
RTECSNumber JG8225000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.