Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, 99+%, for spectroscopy

Catalog No. AC410760010
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle

CAS: 75-12-7 | CH3NO | 45.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2.0°C to 3.0°C
Absorbance 0.01 at 360nm, 0.01 at 400nm, 0.05 at 300nm, 0.2 at 280nm, 0.5 at 275nm
Density 1.1330g/mL
Boiling Point 210.0°C
Quantity 1 L
Flash Point 175°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4460 to 1.4480
Assay Percent Range 99+%
Specific Gravity 1.133
Linear Formula HCONH2
Viscosity 3.75 mPa.s (20°C)
Beilstein 02, 26
Water 0.03% max. (Coulometric)
Fieser 01,402; 02,201; 03,147
Merck Index 15, 4265
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with methanol, ethanol, acetone, acetic, acid, dioxane, ethylene glycol, u.s.p. glycerol,, phenol, slibhtly soluble in ether and benzene
Formula Weight 45.04
Percent Purity ≥99%
Grade Spectroscopy
Chemical Name or Material Formamide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement May damage the unborn child. May cause damage to organs through prolonged or repeated exposure. Suspected of causing cancer if swallowed.
Precautionary Statement GHS P Statement Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Use personal protective equipment as required.
EINECSNumber 200-842-0
RTECSNumber LQ0525000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.