Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Orange G, pure, certified

Catalog No. AC416551000
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle

CAS: 1936-15-8 | C16H10N2Na2O7S2 | 452.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1936-15-8
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.36
MDL Number MFCD00012457
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym Acid Orange 10, 7-Hydroxy-8-(phenylazo)-1, 3-naphthalenedisulfonic acid, disodium salt, C.I. 16230
PubChem CID 131850556
IUPAC Name (8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonic acid;sodium
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O

Specifications

Melting Point 141.0°C
Beilstein 16, 300
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: insoluble in ethanol and diethyl ether
Formula Weight 452.35
Grade Pure
Packaging Glass bottle
Quantity 100 g
Chemical Name or Material Orange G, Pure
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.