Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrabutylammonium Hydroxide Titrant, 0.4M in Water, HPLC Grade
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography, Phase transfer catalyst

Specifications
Specifications
Absorbance | (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.15 max. at 420nm |
Color | Undesignated |
Linear Formula | (CH3(CH2)3)4NOH |
Beilstein | 04, II, 634 |
Concentration | 0.37 to 0.43M |
Fieser | 05,645; 11,500 |
Density | 0.85 |
Solubility Information | Solubility in water: soluble. |
IUPAC Name | tetrabutylazanium;hydroxide |
Formula Weight | 259.47 |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. |
EINECSNumber | 218-147-6 |
RTECSNumber | BS5425000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction