Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium sulfate, 99+%, ACS reagent

Catalog No. AC423400010
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point 280.0°C
Color Silver to Gray
pH 5.0 to 6.0 (5% soln. at 25°C)
Physical Form Granules, Fillings or Chips
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula (NH4)2SO4
Merck Index 15, 552
IUPAC Name sulfuric acid diamine
Formula Weight 132.14
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Ignition 0.005 % max.
Density 1.76g/mL
Chemical Name or Material Ammonium sulfate
Show More Show Less
missing translation for 'healthHazard2' GHS H Statement:
Harmful if swallowed.
missing translation for 'healthHazard3' GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.