Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aniline, 99+%, ACS reagent

Catalog No. AC423420010
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
Packaging:
Glass Bottle

Catalog No. AC423420010

Supplier: Thermo Scientific Chemicals 423420010

Only null left
Add to Cart

CAS: 62-53-3 | C6H7N | 93.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-53-3
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00007629
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
Synonym benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
PubChem CID 6115
ChEBI CHEBI:17296
IUPAC Name aniline
SMILES NC1=CC=CC=C1

Specifications

Melting Point -6.2°C
Color Colorless to White
Density 1.0213g/mL
Boiling Point 181.0°C to 185.0°C
Flash Point 76°C
Ignition Residue 0.005 % max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Refractive Index 1.5845 to 1.5865
Linear Formula C6H5NH2
Quantity 1 L
Beilstein 12, 59
Fieser 06,21; 17,16
Specific Gravity 1.0213
Merck Index 15, 652
Solubility Information Solubility in water: 36g/L (20°C).
Viscosity 4.4 mPa.s (20°C)
Vapor Pressure 0.5mmHg at 20°C
Formula Weight 93.13
Percent Purity 99+%
Grade ACS Reagent
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Aniline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye dam
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 200-539-3
Recommended Storage this product darkens to reddish-brown on storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.