Learn More
Potassium Chloride, 99+%, ACS Reagent

Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | White |
| pH | 5.4 to 8.6 (5% soln. at 25°C) |
| Physical Form | Crystalline Powder and/or Crystals |
| Quantity | 1 kg |
| Assay Percent Range | 99+% |
| Linear Formula | KCl |
| Fieser | 06,480 |
| Merck Index | 15, 7742 |
| IUPAC Name | potassium;chloride |
| Formula Weight | 74.54 |
| Show More |
Safety and Handling
| Hazard Category | HYGROSCOPIC |
| EINECSNumber | 231-211-8 |
| RTECSNumber | TS8050000 |
| TSCA | TSCA |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.