Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium bicarbonate, 99.7+%, ACS reagent

Catalog No. AC424270250
Click to view available options
Quantity:
25 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle

Catalog No. AC424270250

Supplier: Thermo Scientific Chemicals 424270250

Only null left
Add to Cart

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270.0°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.7+%
Linear Formula NaHCO3
Fieser 03,260; 05,595; 11,476; 17,180
Merck Index 15,8719
Solubility Information Solubility in water: 9 g/100mL (20°C). Other solubilities: 16.4 g/100mL water (60°C), insoluble in alcohol
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01
Percent Purity ≥99.7%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium bicarbonate
Show More Show Less
EINECSNumber 205-633-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.