Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH buffer
Specifications
Specifications
| Color | White |
| pH | 8.7 to 9.3 (5% soln. at 25°C) |
| Physical Form | Granular Powder |
| Quantity | 2.5 kg |
| Assay Percent Range | 99+% |
| Linear Formula | Na2HPO4 |
| Merck Index | 15, 8789 |
| Solubility Information | Solubility in water: 75g/L (20°C). Other solubilities: practically insoluble in alcohol |
| IUPAC Name | disodium hydrogen phosphate |
| Formula Weight | 141.96 |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: CAUTION! |
| Precautionary Statement | Emergency Overview May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Hygroscopic. NFPA Health:1 Flammability:0 Instability:1 |
| EINECSNumber | 231-448-7 |
| RTECSNumber | WC4500000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction