Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers

Catalog No. AC428200050
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass bottle
Plastic drum

CAS: 34590-94-8 | C7H16O3 | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34590-94-8
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.2
MDL Number MFCD00059604

Specifications

Melting Point -80.0°C
Density 0.9500g/mL
Boiling Point 180.0°C
Flash Point 75°C
Assay Percent Range 98.5% min. sum of isomers (GC)
Packaging Plastic drum
Linear Formula CH3O(CH2)3O(CH2)3OH
Quantity 5 L
Specific Gravity 0.95
Merck Index 15, 3384
Solubility Information Solubility in water: soluble. Other solubilities: miscible with benzene
Viscosity 4 mPa.s (25°C)
Formula Weight 148.2
Percent Purity 99%
Grade Pure
Chemical Name or Material Di(propylene glycol) methyl ether, Mixture of isomers
Show More Show Less
EINECSNumber 252-104-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.