Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethynylbenzaldehyde, 95%

Catalog No. AC432940010
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Catalog No. AC432940010

Supplier: Thermo Scientific Chemicals 432940010

Only null left
Add to Cart

CAS: 63697-96-1 | C9H6O | 130.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63697-96-1
Molecular Formula C9H6O
Molecular Weight (g/mol) 130.15
InChI Key BGMHQBQFJYJLBP-UHFFFAOYSA-N
PubChem CID 2771645
IUPAC Name 4-ethynylbenzaldehyde
SMILES C#CC1=CC=C(C=C1)C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 89.0°C to 94.5°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Synonym 4-ethynyl benzaldehyde, 4-ethynyl-benzaldehyde, benzaldehyde, 4-ethynyl, benzaldehyde,4-ethynyl, p-ethynylbenzaldehyde, 4-acetylenylbenzaldehyde, acmc-209x0m, 4-ethynylbenzaldehyde, benzaldehyde, 4-ethynyl-9ci
Formula Weight 130.15
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-Ethynylbenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.