Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, specified according to the requirements of Ph.Eur.

Catalog No. AC446270025
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle

CAS: 71-23-8 | C3H8O | 60.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Acidity passes test
Absorbance 0.01 max. at 310nm, 0.02 max. at 290nm, 0.03 max. at 270nm, 0.1 max. at 250nm, 0.3 max. at 230nm
Melting Point -127.0°C
Color Colorless to Yellow
Density 0.8000g/mL
Boiling Point 97.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3840 to 1.3860
Linear Formula CH3CH2CH2OH
Quantity 2.5 L
Merck Index 15,7955
Specific Gravity 0.8
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
Viscosity 2.2 mPa.s (20°C)
Formula Weight 60.1
Percent Purity ≥99.7% (GC)
Related Substances 0.3 (Total impurities) max
Physical Form Liquid
Chemical Name or Material 1-Propanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness.
Causes serious eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
EINECSNumber 200-746-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.