Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl methyl ether, ACS reagent

Catalog No. AC447470010
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy

Catalog No. AC447470010

Supplier: Thermo Scientific Chemicals 447470010

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 1634-04-4 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
IUPAC Name 2-methoxy-2-methylpropane
SMILES COC(C)(C)C

Specifications

Melting Point -110.0°C
Density 0.7404 g/mL
Boiling Point 54.0°C to 56.0°C
Quantity 1 L
Flash Point -28°C
Packaging Glass Bottle
Refractive Index 1.3675 to 1.3695
Specific Gravity 0.7404
Linear Formula (CH3)3COCH3
Viscosity 0.36 mPa.s (20°C)
Merck Index 15, 6106
Solubility Information Solubility aq. soln.: 51 g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5 g/100 g
Formula Weight 88.15
Grade ACS Reagent
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes skin irritation.

Highly flammable liquid and vapor.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

Ground and bond container and receiving equipment
EINECSNumber 216-653-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.