Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrogen peroxide, 12% w/v
Click to view available options
Quantity:
100 mL
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 100.0°C |
| Density | 1.0500 g/mL |
| Quantity | 100 mL |
| Molecular Formula | H2O2 |
| Linear Formula | H2O2 |
| Specific Gravity | 1.05 |
| Solubility Information | Solubility aq. soln.: soluble. |
| Formula Weight | 34.01 |
| Chemical Name or Material | Hydrogen peroxide |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May intensify fire; oxidizer. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wash face, hands and any exposed skin thoroughly after handling. Do not eat, drink or smoke when using this product. |
| EINECSNumber | 231-765-0 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction