Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid, 98%, Thermo Scientific™

Catalog No. AC471670010
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym TFA
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15°C
Boiling Point 72°C (760 mmHg)
Appearance Colorless to light yellow clear liquid
Linear Formula CF3CO2H
UN Number 2699
Solubility Information Miscible in water, miscible with ether, acetone, ethanol, benzene, CCl4, hexane, methanol, xylene
Concentration 97.5% to 102.5%
Formula Weight 114.02
Percent Purity 98%
Grade Pure
Density 1.535
Quantity 1 kg
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3
Sensitivity Hygroscopic
Vapor Pressure 107 mbar (25°C)
Chemical Name or Material Trifluoroacetic acid
Show More Show Less
Hazard Category GHS07: Exclamation mark
Hazard Statement GHS05: Corrosion
ShelfLife 2 years
RTECSNumber AJ9625000
Recommended Storage Normal conditions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.