Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trifluoroacetic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Specifications
Specifications
Melting Point | -15°C |
Boiling Point | 72°C (760 mmHg) |
Appearance | Colorless to light yellow clear liquid |
Linear Formula | CF3CO2H |
UN Number | 2699 |
Solubility Information | Miscible in water, miscible with ether, acetone, ethanol, benzene, CCl4, hexane, methanol, xylene |
Concentration | 97.5% to 102.5% |
Formula Weight | 114.02 |
Percent Purity | 98% |
Grade | Pure |
Show More |
Safety and Handling
Hazard Category | GHS07: Exclamation mark |
Hazard Statement | GHS05: Corrosion |
ShelfLife | 2 years |
EINECSNumber | 200-929-3 |
RTECSNumber | AJ9625000 |
Recommended Storage | Normal conditions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction