Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Aqualine™ Electrolyte A, for Karl Fischer Titration By Coulometry, Fisher Chemical

Catalog No. AL2500500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy

Catalog No. AL-2500500

Only null left
Add to Cart
This item is not returnable. View return policy

Anolyte solution for general use in conventional cells with a diaphragm

  • Anolyte solution for use in conventional cells with a diaphragm
  • Source of electrolyte required for anodic oxidation for coulometric analysis
  • Contains methanol and chloroform as solvents
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aqualine™ Electrolyte A
Name Note Karl Fischer Reagent
Color Colorless
CAS 108-75-8, 7446-09-5, 67-66-3, 111-42-2
Physical Form Liquid
Quantity 500 mL
Packaging Amber glass bottle
MDL Number 81734
UN Number 1992
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.