Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Sulfate (Powder/USP), Fisher Chemical™

Catalog No. B75500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail

Catalog No. B75-500

Only null left
Add to Cart

Fine white powder, free from grittiness

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
IUPAC Name barium(2+) sulfate
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1580°C
Color Yellow
Physical Form Solid
Quantity 500 g
Assay Percent Range 97.5 to 100.5 %
Formula Weight 233.39
Percent Purity 97.5 to 100.5%
Grade USP
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Barium Sulfate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.