Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Agarose (High-EEO/Protein Electrophoresis Grade), Fisher BioReagents
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
Description
Immunoelectrophoresis (IEP). Crossed-IEP. Counter-immunoelectrophoresis (CIEP). Serum protein electrophoresis.

Specifications
Specifications
Physical Form | Powder/Solid |
Color | Cream |
Quantity | 100 g |
DNase | DNase free |
Electroendosmosis (EEO) | 0.23 to 0.26 |
Gelation Temperature | 34.5° to 37.5°C |
Gel Strength | 650g/cm2min. |
Packaging | Poly Bottle |
Formula Weight | 306.12 |
Grade | Protein Electrophoresis |
Show More |
Safety and Handling

Recommended Storage | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction