Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail
Description
For protein extraction procedures and SDS polyacrylamide gels
- DNase and RNase-free detergent for protein extraction procedures and polyacrylamide gel electrophoresis
- SDS is a commonly used detergent in protein purification and polyacrylamide gel electrophoresis (PAGE)
- SDS denatures proteins by disrupting the non-covalent bonds of the protein
- Functionally tested for use in electrophoresis
- Intended for research use only; not for human or animal diagnostic purposes

Specifications
Specifications
Absorbance | 0.1 max. (0.1% solution) at 280nm, 0.4 max. (0.1% solution) at 230nm |
Color | White |
CAS | 151-21-3 |
Physical Form | Solid |
Melting Point | 206°C |
pH | 8.5 to 10 |
Quantity | 5 kg |
Identification | Pass Test |
Packaging | Poly Pail |
Assay Percent Range | (as Fatty Alcohol Sulfate) >=99% |
Show More |
Safety and Handling

Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Flammable solid. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory reaction. Avoid contact with skin and eyes. Do not breathe dust. Use explosion-proof equipment. Use only non-sparking tools. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Remove from exposure, lie down. Move to fresh air. NFPA Health:2 Flammability:3 Instability:0 |
Recommended Storage | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction