Learn More
Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™
Shop All Fisherbrand Products

Description
Phenol, Saturated (pH 6.6/7.9, Liq.)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Assay Percent Range | ≥99.0 |
| UN Number | 2821 |
| Percent Purity | ≥99.0% |
| Identification | Pass Test |
| Packaging | Coated Amber Glass/Poison Pack |
| Quantity | 400 mL |
| Physical Form | Liquid |
| Chemical Name or Material | Phenol, Saturated |
| Grade | DNase- and RNase-Free |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Combustible liquid. Causes burns by all exposure routes. Possible risks of irreversible effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. NFPA Health:3 Flammability:2 Instability:0 |
| DOTInformation | DOT Class 6.1, : Poison |
| Recommended Storage | 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.