Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formamide (Molecular Biology), Fisher BioReagents
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Description
Molecular Biology-grade Formamide is commonly used in nucleic acid hybridization and sequencing. It requires pretreatment with a mixed-bed resin.

Specifications
Specifications
pH | 4 to 5 |
Melting Point | 2°C |
Absorbance | 0.1 max. at 280nm |
Density | 1.133g/cm³ |
Boiling Point | 210°C |
Quantity | 100 mL |
Identification | Pass Test |
Packaging | Glass Bottle |
Physical Form | Liquid |
Assay Percent Range | ≥99 % |
Show More |
Safety and Handling

Recommended Storage | 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction