Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polyethylene Glycol 8000 (PEG), Fisher BioReagents™
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Description
Polyethylene Glycol 8000, HO-[CH2CH2O]n-H, CAS Number-25322-68-3, Polyethylene Oxide, Carbowax, PEG 8000, 1kg, Gray, White, 7000 to 9000, CAUTION!
PEG 8000 is suitable as a medium for the fusion of mammalian cells. It is also used to precipitate bacteriophage from lysed cell supernatants.
Specifications
Specifications
| Color | White |
| pH | 5.0 to 7.0 |
| Physical Form | Solid |
| Melting Point | 55°C |
| Linear Formula | H(OCH2CH2)nOH |
| Merck Index | 15, 7688 |
| Quantity | 100 g |
| Formula Weight | 7000 to 9000 |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:1 Flammability:1 Instability:0 |
| Recommended Storage | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction